Thermo Scientific Chemicals

N-Ethyldiisopropylamine, 99%, Thermo Scientific Chemicals

Catalog number: A11801.AE
100 mL, Each
Thermo Scientific Chemicals

N-Ethyldiisopropylamine, 99%, Thermo Scientific Chemicals

Catalog number: A11801.AE
100 mL, Each
Quantity
Catalog number: A11801.AE
also known as A11801-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7087-68-5
IUPAC Name
ethylbis(propan-2-yl)amine
Molecular Formula
C8H19N
InChI Key
JGFZNNIVVJXRND-UHFFFAOYSA-N
SMILES
CCN(C(C)C)C(C)C
Assay (GC)
≥98.5% (non-U.S. specification)
Water Content (Karl Fischer Titration)
≤0.2% (U.S. specification)
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms (non-U.S. specification)
Refractive Index
1.4120-1.4150 @ 20°C (non-U.S. specification)

Description

N-Ethyldiisopropylamine is used to make or formulate industrial products. It is also used in organic chemistry as a base. It acts as a selective reagent in the alkylation of secondary amines to tertiary amines by alkyl halides. It reacts with monoactivated Michael acceptors to afford symmetrical sulfones. Further, it is used to prepare heterocyclic compound, sscorpionine by a reaction with disulfur dichloride using 1,4-diazabicyclo[2.2.2]octane (DABCO) as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Ethyldiisopropylamine is used to make or formulate industrial products. It is also used in organic chemistry as a base. It acts as a selective reagent in the alkylation of secondary amines to tertiary amines by alkyl halides. It reacts with monoactivated Michael acceptors to afford symmetrical sulfones. Further, it is used to prepare heterocyclic compound, sscorpionine by a reaction with disulfur dichloride using 1,4-diazabicyclo[2.2.2]octane (DABCO) as a catalyst.

Solubility
Miscible with water and ethanol.

Notes
Incompatible with acids, acid chlorides, acid anhydrides, carbon dioxide, copper, brass, rubber, oxidizing agents, nitrates, peroxides, aldehydes and metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text