Manganese(II) sulfate monohydrate, 97%, Thermo Scientific Chemicals
Manganese(II) sulfate monohydrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manganese(II) sulfate monohydrate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
500 g
Catalog number A11809.0E
also known as A11809-0E
Price (USD)/ Each
140.65
Online exclusive
156.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
140.65
Online exclusive
156.00 
Save 15.35 (10%)
Add to cart
Manganese(II) sulfate monohydrate, 97%, Thermo Scientific Chemicals
Catalog numberA11809.0E
Price (USD)/ Each
140.65
Online exclusive
156.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10034-96-5
IUPAC Namemanganese(2+) hydrate sulfate
Molecular FormulaH2MnO5S
InChI KeyISPYRSDWRDQNSW-UHFFFAOYSA-L
SMILESO.[Mn++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale pink to pink
FormCrystals or powder or crystalline powder
Complexometric Titration≥96.0 to ≤104.0%
Manganese(II) sulfate is used as a colorant in dyes. It is also used as feed additives, catalyst for the synthesis of chlorophyll, make plants grow well and increase production. It is used for making dryer of ink and paint. It is used in paper making , porcelain/ceramic, medicine and other industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(II) sulfate is used as a colorant in dyes. It is also used as feed additives, catalyst for the synthesis of chlorophyll, make plants grow well and increase production. It is used for making dryer of ink and paint. It is used in paper making , porcelain/ceramic, medicine and other industries.

Solubility
Soluble in water. 5-10 g/100 mL at 21°C

Notes
Loses all water at 400-450°
RUO – Research Use Only

General References:

  1. Joesph A. Rard. Isopiestic determination of the osmotic and activity coefficients of aqueous manganese(II) chloride, manganese(II) sulfate, and rubidium chloride at 25 .degree.C J. Chem. Eng. Data.1984, 29 (4), 443-450.
  2. Ali Ramazaniab; Ali Reza Kazemizadehb & Farzin Marandib. Phosphorus, Sulfur, and Silicon and the Related Elements. 2007, 180 1541-1544.