Thermo Scientific Chemicals

Manganese(II) sulfate monohydrate, 97%, Thermo Scientific Chemicals

Catalog number: A11809.36
500 g, Each
Thermo Scientific Chemicals

Manganese(II) sulfate monohydrate, 97%, Thermo Scientific Chemicals

Catalog number: A11809.36
500 g, Each
Quantity
Catalog number: A11809.36
also known as A11809-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10034-96-5
IUPAC Name
manganese(2+) hydrate sulfate
Molecular Formula
H2MnO5S
InChI Key
ISPYRSDWRDQNSW-UHFFFAOYSA-L
SMILES
O.[Mn++].[O-]S([O-])(=O)=O
Appearance (Color)
Pale pink to pink
Form
Crystals or powder or crystalline powder
Complexometric Titration
≥96.0 to ≤104.0%

Description

Manganese(II) sulfate is used as a colorant in dyes. It is also used as feed additives, catalyst for the synthesis of chlorophyll, make plants grow well and increase production. It is used for making dryer of ink and paint. It is used in paper making , porcelain/ceramic, medicine and other industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(II) sulfate is used as a colorant in dyes. It is also used as feed additives, catalyst for the synthesis of chlorophyll, make plants grow well and increase production. It is used for making dryer of ink and paint. It is used in paper making , porcelain/ceramic, medicine and other industries.

Solubility
Soluble in water. 5-10 g/100 mL at 21°C

Notes
Loses all water at 400-450°
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text