Thermo Scientific Chemicals

2-Methylglutaric acid, 98%, Thermo Scientific Chemicals

Catalog number: A11823.22
100 g, Each
Thermo Scientific Chemicals

2-Methylglutaric acid, 98%, Thermo Scientific Chemicals

Catalog number: A11823.22
100 g, Each
Quantity
Catalog number: A11823.22
also known as A11823-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
617-62-9
IUPAC Name
2-methylpentanedioic acid
Molecular Formula
C6H10O4
InChI Key
AQYCMVICBNBXNA-UHFFFAOYNA-N
SMILES
CC(CCC(O)=O)C(O)=O
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (Silylated GC)
≥97.5%
Identification (FTIR)
Conforms

Description

2-Methylglutaric acid is used as primary and secondary intermediates in pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylglutaric acid is used as primary and secondary intermediates in pharmaceutical and chemical research.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Incompatible with bases and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text