Thermo Scientific Chemicals

4-Bromoanisole, 99%, Thermo Scientific Chemicals

Catalog number: A11824.36
500 g, Each
Thermo Scientific Chemicals

4-Bromoanisole, 99%, Thermo Scientific Chemicals

Catalog number: A11824.36
500 g, Each
Quantity
Catalog number: A11824.36
also known as A11824-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
104-92-7
IUPAC Name
1-bromo-4-methoxybenzene
Molecular Formula
C7H7BrO
InChI Key
QJPJQTDYNZXKQF-UHFFFAOYSA-N
SMILES
COC1=CC=C(Br)C=C1
Form
Liquid
Identification (FTIR)
Conforms
Assay (GC)
≥98.5%
Appearance (Color)
Clear colorless to very pale yellow
Refractive Index
1.5625-1.5655 @ 20?C

Description

4-Bromoanisole finds application as an intermediate in synthetic chemistry. It is used in the preparation of aryl 1,3-diketones and ethyl 4-methoxycinnamate. It is a used as a brominating reagent. Further, it is used in Suzuki coupling reaction with phenylboronic acid as well as in Heck reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromoanisole finds application as an intermediate in synthetic chemistry. It is used in the preparation of aryl 1,3-diketones and ethyl 4-methoxycinnamate. It is a used as a brominating reagent. Further, it is used in Suzuki coupling reaction with phenylboronic acid as well as in Heck reaction.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text