Methyl 3-(bromomethyl)benzoate is used in organic synthesis of other chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 3-(bromomethyl)benzoate is used in organic synthesis of other chemicals.
Solubility
Slightly soluble in water.
Notes
Store at 4°C. Keep away from strong bases and oxidizing agents. Light sensitive. Incompatible with reducing agents, amines.
RUO – Research Use Only
Amanda R. Knapp; Matthew J. Panzner; Doug A. Medvetz; Brian D. Wright; Claire A. Tessier; Wiley J. Youngs. Synthesis and antimicrobial studies of silver N-heterocyclic carbene complexes bearing a methyl benzoate substituent.Inorganica chimica Acta.2010, 364 125-131.