2,4,6-Trimethylbenzoic acid, 99%, Thermo Scientific Chemicals
2,4,6-Trimethylbenzoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4,6-Trimethylbenzoic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A11831.09
also known as A11831-09
Price (USD)/ Each
68.65
Online exclusive
76.50 
Save 7.85 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
68.65
Online exclusive
76.50 
Save 7.85 (10%)
Add to cart
2,4,6-Trimethylbenzoic acid, 99%, Thermo Scientific Chemicals
Catalog numberA11831.09
Price (USD)/ Each
68.65
Online exclusive
76.50 
Save 7.85 (10%)
-
Add to cart
Chemical Identifiers
CAS480-63-7
IUPAC Name2,4,6-trimethylbenzoic acid
Molecular FormulaC10H12O2
InChI KeyFFFIRKXTFQCCKJ-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(C(O)=O)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Identification (FTIR)Conforms
Assay (Silylated GC)≥98.5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only