3-Iodobenzotrifluoride, 98+%, Thermo Scientific Chemicals
3-Iodobenzotrifluoride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Iodobenzotrifluoride, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number A11843.18
also known as A11843-18
Price (USD)/ Each
87.30
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
87.30
Add to cart
3-Iodobenzotrifluoride, 98+%, Thermo Scientific Chemicals
Catalog numberA11843.18
Price (USD)/ Each
87.30
-
Add to cart
Chemical Identifiers
CAS401-81-0
IUPAC Name1-iodo-3-(trifluoromethyl)benzene
Molecular FormulaC7H4F3I
InChI KeyIGISPMBUGPHLBY-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CC(I)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Refractive Index1.5165-1.5205 @ 20°C
Appearance (Color)Clear colorless to pale yellow or pale pink
FormLiquid
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only