Thermo Scientific Chemicals

2,2,3,3,3-Pentafluoro-1-propanol, 98%, Thermo Scientific Chemicals

Catalog number: A11846.14
25 g, Each
Thermo Scientific Chemicals

2,2,3,3,3-Pentafluoro-1-propanol, 98%, Thermo Scientific Chemicals

Catalog number: A11846.14
25 g, Each
Quantity
Catalog number: A11846.14
also known as A11846-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
422-05-9
IUPAC Name
2,2,3,3,3-pentafluoropropan-1-ol
Molecular Formula
C3H3F5O
InChI Key
PSQZJKGXDGNDFP-UHFFFAOYSA-N
SMILES
OCC(F)(F)C(F)(F)F
Assay from Supplier's CofA
≥97.5% (US-sourced material)
Comment
Material sourced in UK and US
Identification (FTIR)
Conforms
Identification (FTIR)
Conforms (US-sourced material)
Appearance (Color)
Clear colorless

Description

2,2,3,3,3-Pentafluoro-1-propanol is used as Pharmaceutical Intermediates. It can be used in agrochemical, pharmaceutical and dyestuff field etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3,3-Pentafluoro-1-propanol is used as Pharmaceutical Intermediates. It can be used in agrochemical, pharmaceutical and dyestuff field etc.

Solubility
Soluble in water.21.9 g/L at 25°C

Notes
Incompatible with strong oxidizing agents, Acid chlorides, Acid anhydrides. Keep away from heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text