Thermo Scientific Chemicals

Hexamethyldisiloxane, 98+%, Thermo Scientific Chemicals

Catalog number: A11848.AE
100 mL, Each
Thermo Scientific Chemicals

Hexamethyldisiloxane, 98+%, Thermo Scientific Chemicals

Catalog number: A11848.AE
100 mL, Each
Quantity
Catalog number: A11848.AE
also known as A11848-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-46-0
IUPAC Name
trimethyl[(trimethylsilyl)oxy]silane
Molecular Formula
C6H18OSi2
InChI Key
UQEAIHBTYFGYIE-UHFFFAOYSA-N
SMILES
C[Si](C)(C)O[Si](C)(C)C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.0%
Refractive Index
1.3760-1.3800 @ 20?C
Identification (FTIR)
Conforms

Description

Hexamethyldisiloxane is used as a solvent in organic reactions. It finds application as an international standard for 1H NMR spectroscopy for calibrating chemical shift. Further, it is used in liquid bandages to protect skin from irritation. In addition, it is used to measure tissue oxygen tension as a receptor molecule. It is also used to make low-k dielectric materials for the semi-conductor industries by plasma-enhanced chemical vapor deposition(PECVD) method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethyldisiloxane is used as a solvent in organic reactions. It finds application as an international standard for 1H NMR spectroscopy for calibrating chemical shift. Further, it is used in liquid bandages to protect skin from irritation. In addition, it is used to measure tissue oxygen tension as a receptor molecule. It is also used to make low-k dielectric materials for the semi-conductor industries by plasma-enhanced chemical vapor deposition(PECVD) method.

Solubility
Slightly miscible with water.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, bases and oxygen.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text