Thermo Scientific Chemicals

trans-4-Hydroxy-L-proline, 99+%, Thermo Scientific Chemicals

Catalog number: A11851.22
100 g, Each
Thermo Scientific Chemicals

trans-4-Hydroxy-L-proline, 99+%, Thermo Scientific Chemicals

Catalog number: A11851.22
100 g, Each
Quantity
Catalog number: A11851.22
also known as A11851-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
51-35-4
IUPAC Name
4-hydroxypyrrolidine-2-carboxylic acid
Molecular Formula
C5H9NO3
InChI Key
PMMYEEVYMWASQN-UHFFFAOYNA-N
SMILES
OC1CNC(C1)C(O)=O
Appearance (Color)
White to pale cream
Optical Rotation
-75.6 ± 0.5° (C=4 in water)
Assay (Non-aqueous acid-base Titration)
≥99.0 to ≤101.0%
Identification (FTIR)
Conforms (non-U.S. specification)
Form
Crystals or powder or crystalline powder

Description

trans-4-Hydroxy-L-proline is used as a reagent for the preparation of neuroexcitatory kainoids and antifungal echinocandins. It is also involved in the synthesis of chiral ligands for enantioselective ethylation of aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-Hydroxy-L-proline is used as a reagent for the preparation of neuroexcitatory kainoids and antifungal echinocandins. It is also involved in the synthesis of chiral ligands for enantioselective ethylation of aldehydes.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text