Thermo Scientific Chemicals

4-tert-Butylbenzyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: A11856.14
25 g, Each
Thermo Scientific Chemicals

4-tert-Butylbenzyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: A11856.14
25 g, Each
Quantity
Catalog number: A11856.14
also known as A11856-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19692-45-6
IUPAC Name
1-tert-butyl-4-(chloromethyl)benzene
Molecular Formula
C11H15Cl
InChI Key
WAXIFMGAKWIFDQ-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC=C(CCl)C=C1
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%
Refractive Index
1.5190-1.5220 @ 20?C

Description

4-tert-Butylbenzyl chloride has been used to control molecular weight and prevent microgel formation during polymerization reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylbenzyl chloride has been used to control molecular weight and prevent microgel formation during polymerization reaction.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent. Store away from strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text