Thermo Scientific Chemicals

Theobromine, 99%, Thermo Scientific Chemicals

Catalog number: A11861.30
250 g, Each
Thermo Scientific Chemicals

Theobromine, 99%, Thermo Scientific Chemicals

Catalog number: A11861.30
250 g, Each
Quantity
Catalog number: A11861.30
also known as A11861-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
83-67-0
IUPAC Name
3,7-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular Formula
C7H8N4O2
InChI Key
YAPQBXQYLJRXSA-UHFFFAOYSA-N
SMILES
CN1C=NC2=C1C(=O)NC(=O)N2C
Identification (FTIR)
Conforms
Form
Powder
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5% (on dried substance)
Loss on Drying
≤0.5% (1g, 105?C)

Description

Phosphodiesterase inhibitor, weak adenosine receptor antagonist, diuretic and smooth muscle relaxant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphodiesterase inhibitor, weak adenosine receptor antagonist, diuretic and smooth muscle relaxant.

Solubility
Soluble in hot methanol and dimethyl sulfoxide.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent. Store away from strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text