Thermo Scientific Chemicals

Sodium thiocyanate, 96%, Thermo Scientific Chemicals

Catalog number: A11867.0B
1000 g, Each
Thermo Scientific Chemicals

Sodium thiocyanate, 96%, Thermo Scientific Chemicals

Catalog number: A11867.0B
1000 g, Each
Quantity
Catalog number: A11867.0B
also known as A11867-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
540-72-7
IUPAC Name
sodium cyanosulfanide
Molecular Formula
CNNaS
InChI Key
VGTPCRGMBIAPIM-UHFFFAOYSA-M
SMILES
[Na+].[S-]C#N
Form
Crystals or powder or crystalline powder or fused/lumpy solid or chunks
Assay (Argentometric Titration)
≥95.0 to ≤105.0%
Water Content (Karl Fischer Titration)
≤5.0%
Appearance (Color)
White

Description

A compound used to convert alkyl halides into the corresponding alkylthiocyanates. Undergoes nucleophilic substitution reactions with phase-transfer catalysts. Silver thiocyanate may be used as well, the precipitation of insoluble silver halides help simplify workup. Treatment of isopropyl bromide with sodium thiocyanate in a hot ethanolic solution affords isopropyl thiocyanate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A compound used to convert alkyl halides into the corresponding alkylthiocyanates. Undergoes nucleophilic substitution reactions with phase-transfer catalysts. Silver thiocyanate may be used as well, the precipitation of insoluble silver halides help simplify workup. Treatment of isopropyl bromide with sodium thiocyanate in a hot ethanolic solution affords isopropyl thiocyanate.

Solubility
Soluble in water (1250g/L), alcohol, and acetone.

Notes
Hygroscopic, protect from humidity and water. Store in cool, dry conditions in well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text