FormCrystals or powder or crystalline powder
Assay (GC)≥98.0% (non-U.S. specification)
Assay from Suppliers CofA≥98.0% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Melting Point (clear melt)96-108°C
View more