It employed as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It employed as a intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
RUO – Research Use Only
Luca Bruche; Maria Luisa Gelmi; Gaetano Zecchi. 1,3-Dipolar cycloadditions of 3,5-dichloro-2,4,6-trimethylbenzonitrile oxide to (phenylsulfonyl)allenes. J. Org. Chem,1985, 50(17), 3206-3208.
W.E. Acree Jr.; Sheryl A. Tucker; Anita I. Zvaigzne; Yang Meng-Yan; G. Pilcher; Maria D.M.C. Ribeiro Da Silva. Enthalpies of combustion of 2,4,6-trimethylbenzonitrile, 2,4,6-trimethylbenzonitrile N-oxide, 2,6-dimethylbenzonitrile, 2,4,6-trimethoxybenzonitrile, and 2,4,6-trimethoxybenzonitrile N-oxide: the dissociation enthalpies of the (Nsingle bondO) bonds. The Journal of Chemical Thermodynamics,1991, 23(1), 31-36.