Thermo Scientific Chemicals

3-Bromo-2-methylaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A11884.14
25 g, Each
Thermo Scientific Chemicals

3-Bromo-2-methylaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A11884.14
25 g, Each
Quantity
Catalog number: A11884.14
also known as A11884-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
55289-36-6
IUPAC Name
3-bromo-2-methylaniline
Molecular Formula
C7H8BrN
InChI Key
IILVSKMKMOJHMA-UHFFFAOYSA-N
SMILES
CC1=C(N)C=CC=C1Br
Appearance (Color)
Clear colorless to brown or orange
Refractive Index
1.6125-1.6195 @ 20?C
Assay (GC)
≥98.0%
Form
Liquid
Identification (FTIR)
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text