Formamidinesulfinic acid, 98%, Thermo Scientific Chemicals
Formamidinesulfinic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Formamidinesulfinic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A11885.22
also known as A11885-22
Price (USD)/ Each
30.65
Online exclusive
34.10 
Save 3.45 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
30.65
Online exclusive
34.10 
Save 3.45 (10%)
Add to cart
Formamidinesulfinic acid, 98%, Thermo Scientific Chemicals
Catalog numberA11885.22
Price (USD)/ Each
30.65
Online exclusive
34.10 
Save 3.45 (10%)
-
Add to cart
Chemical Identifiers
CAS1758-73-2
IUPAC Namecarbamimidoylsulfinic acid
Molecular FormulaCH4N2O2S
InChI KeyFYOWZTWVYZOZSI-UHFFFAOYNA-N
SMILESNC(=N)S(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Appearance (Color)White to pale yellow
Assay (unspecified)≥ 97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only