Thermo Scientific Chemicals

Formamidinesulfinic acid, 98%, Thermo Scientific Chemicals

Catalog number: A11885.36
500 g, Each
Thermo Scientific Chemicals

Formamidinesulfinic acid, 98%, Thermo Scientific Chemicals

Catalog number: A11885.36
500 g, Each
Quantity
Catalog number: A11885.36
also known as A11885-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1758-73-2
IUPAC Name
carbamimidoylsulfinic acid
Molecular Formula
CH4N2O2S
InChI Key
FYOWZTWVYZOZSI-UHFFFAOYNA-N
SMILES
NC(=N)S(O)=O
Form
Crystalline powder
Appearance (Color)
White to pale yellow
Assay (unspecified)
≥ 97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only