Thermo Scientific Chemicals

Michler's Ketone, 98%, Thermo Scientific Chemicals

Catalog number: A11889.36
500 g, Each
Thermo Scientific Chemicals

Michler's Ketone, 98%, Thermo Scientific Chemicals

Catalog number: A11889.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A11889.36
also known as A11889-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
90-94-8
IUPAC Name
4-[4-(dimethylamino)benzoyl]-N,N-dimethylaniline
Molecular Formula
C17H20N2O
InChI Key
VVBLNCFGVYUYGU-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)N(C)C
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥97.5%
Appearance (Color)
Pale blue to dark blue to dark gray

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text