Thermo Scientific Chemicals

1,4-Dichloro-2-nitrobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A11896.36
500 g, Each
Thermo Scientific Chemicals

1,4-Dichloro-2-nitrobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A11896.36
500 g, Each
Quantity
Catalog number: A11896.36
also known as A11896-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
89-61-2
IUPAC Name
1,4-dichloro-2-nitrobenzene
Molecular Formula
C6H3Cl2NO2
InChI Key
RZKKOBGFCAHLCZ-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC(Cl)=CC=C1Cl
Appearance (Color)
Pale yellow to yellow
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder or lumps
Melting Point (clear melt)
52.0-58.0?C

Description

1,4-Dichloro-2-nitrobenzene is a reagent used in the synthesis of antitrypanosomal, antileishmanial and antimalarial agents. It is also used in the synthesis of lysophosphatidic acid acyltransferase-β in hibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dichloro-2-nitrobenzene is a reagent used in the synthesis of antitrypanosomal, antileishmanial and antimalarial agents. It is also used in the synthesis of lysophosphatidic acid acyltransferase-β in hibitors.

Solubility
Soluble in water, ethanol, ether, benzene, carbon disulfide. Slightly soluble in carbon tetrachloride.

Notes
Incompatible with strong oxidizing agents, strong bases.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text