6,7-Dihydroxy-4-methylcoumarin, 97%, Thermo Scientific Chemicals
6,7-Dihydroxy-4-methylcoumarin, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6,7-Dihydroxy-4-methylcoumarin, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number A11902.14
also known as A11902-14
Price (USD)/ Each
100.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
100.00
Add to cart
6,7-Dihydroxy-4-methylcoumarin, 97%, Thermo Scientific Chemicals
Catalog numberA11902.14
Price (USD)/ Each
100.00
-
Add to cart
Chemical Identifiers
CAS529-84-0
IUPAC Name6,7-dihydroxy-4-methyl-2H-chromen-2-one
Molecular FormulaC10H8O4
InChI KeyKVOJTUXGYQVLAJ-UHFFFAOYSA-N
SMILESCC1=CC(=O)OC2=CC(O)=C(O)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
Appearance (Color)Pale brown or grey
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only