Thermo Scientific Chemicals

Bromoform, 96%, stab. with ethanol, Thermo Scientific Chemicals

Catalog number: A11904.0B
1000 g, Each
Thermo Scientific Chemicals

Bromoform, 96%, stab. with ethanol, Thermo Scientific Chemicals

Catalog number: A11904.0B
1000 g, Each
Quantity
Catalog number: A11904.0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-25-2
IUPAC Name
tribromomethane
Molecular Formula
CHBr3
InChI Key
DIKBFYAXUHHXCS-UHFFFAOYSA-N
SMILES
BrC(Br)Br
Form
Liquid
Stabilizer
Ethanol 1-5%
Assay (GC)
≥95.0%
Appearance (Color)
Clear colorless to yellow to orange
Identification (FTIR)
Conforms

Description

Bromoform is widely used as a solvent for waxes, oils and greases. It is utilized for mineral ore separation in geological tests. It is used as an intermediate in chemical synthesis as well as a laboratory reagent. It is the ingredient of fire-resistant chemicals and fluid gauges. It acts as a sedative and as cough reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromoform is widely used as a solvent for waxes, oils and greases. It is utilized for mineral ore separation in geological tests. It is used as an intermediate in chemical synthesis as well as a laboratory reagent. It is the ingredient of fire-resistant chemicals and fluid gauges. It acts as a sedative and as cough reducing agent.

Solubility
Slightly soluble in water.

Notes
Light-sensitive. A lachrymator. Incompatible with chemically active metals and strong bases.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text