Thermo Scientific Chemicals

4-Ethylmorpholine, 98%, Thermo Scientific Chemicals

Catalog number: A11905.AU
1000 mL, Each
Thermo Scientific Chemicals

4-Ethylmorpholine, 98%, Thermo Scientific Chemicals

Catalog number: A11905.AU
1000 mL, Each
Quantity
Catalog number: A11905.AU
also known as A11905-AU
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-74-3
IUPAC Name
4-ethylmorpholine
Molecular Formula
C6H13NO
InChI Key
HVCNXQOWACZAFN-UHFFFAOYSA-N
SMILES
CCN1CCOCC1
Identification (FTIR)
Conforms
Refractive Index
1.4395-1.4435 @ 20°C
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥97.5%

Description

N-Ethylmorpholine is a component of the buffer used in basic peptide separation through anion-exchange chromatography. Further, it acts as a catalyst in the preparation of polyurethane foam.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Ethylmorpholine is a component of the buffer used in basic peptide separation through anion-exchange chromatography. Further, it acts as a catalyst in the preparation of polyurethane foam.

Solubility
Miscible with water.

Notes
Light and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text