Thermo Scientific Chemicals

1-Iodododecane, 98%, stab. with copper, Thermo Scientific Chemicals

Catalog number: A11909.22
100 g, Each
Thermo Scientific Chemicals

1-Iodododecane, 98%, stab. with copper, Thermo Scientific Chemicals

Catalog number: A11909.22
100 g, Each
Quantity
Catalog number: A11909.22
also known as A11909-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4292-19-7
IUPAC Name
1-iodododecane
Molecular Formula
C12H25I
InChI Key
GCDPERPXPREHJF-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCI
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow to pale orange
Assay (GC)
≥97.5%
Refractive Index
1.4825-1.4860 @ 20?C
Form
Liquid

Description

1-Iodododecane has been used in the preparation of dodecylated graphite, alkyl functionalized single walled carbon nanotubes (SWNTs). It is also used as chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodododecane has been used in the preparation of dodecylated graphite, alkyl functionalized single walled carbon nanotubes (SWNTs). It is also used as chemical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Light Sensitive. Keep away from oxidizing agents. Store in dark.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text