Methyl vinyl ketone, tech. 90%, stab., Thermo Scientific Chemicals
Methyl vinyl ketone, tech. 90%, stab., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl vinyl ketone, tech. 90%, stab., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A11910.AC
also known as A11910-AC
Price (USD)/ Each
34.65
Online exclusive
38.10 
Save 3.45 (9%)
-
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (USD)/ Each
34.65
Online exclusive
38.10 
Save 3.45 (9%)
Add to cart
Methyl vinyl ketone, tech. 90%, stab., Thermo Scientific Chemicals
Catalog numberA11910.AC
Price (USD)/ Each
34.65
Online exclusive
38.10 
Save 3.45 (9%)
-
Add to cart
Chemical Identifiers
CAS78-94-4
IUPAC Namebut-3-en-2-one
Molecular FormulaC4H6O
InChI KeyFUSUHKVFWTUUBE-UHFFFAOYSA-N
SMILESCC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange
FormLiquid
Assay (GC)≥80.0%
CommentMay darken on storage
Stabilizer≤0.7% Acetonitrile
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only