4-Chlorobenzhydrazide, 98%, Thermo Scientific Chemicals
4-Chlorobenzhydrazide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chlorobenzhydrazide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
100 g
Catalog number A11911.18
also known as A11911-18
Price (USD)/ Each
184.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
184.00
Add to cart
4-Chlorobenzhydrazide, 98%, Thermo Scientific Chemicals
Catalog numberA11911.18
Price (USD)/ Each
184.00
-
Add to cart
Chemical Identifiers
CAS536-40-3
IUPAC Name4-chlorobenzohydrazide
Molecular FormulaC7H7ClN2O
InChI KeyPKBGHORNUFQAAW-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White to pale cream
Melting Point (clear melt)162.0-168.0?C
FormCrystals or powder or crystalline powder or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only