Thermo Scientific Chemicals

2-Amino-5-bromobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A11915.06
5 g, Each
Thermo Scientific Chemicals

2-Amino-5-bromobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A11915.06
5 g, Each
Quantity
Catalog number: A11915.06
also known as A11915-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5794-88-7
IUPAC Name
2-amino-5-bromobenzoic acid
Molecular Formula
C7H6BrNO2
InChI Key
CUKXRHLWPSBCTI-UHFFFAOYSA-N
SMILES
NC1=CC=C(Br)C=C1C(O)=O
Appearance (Color)
Pale cream to cream to pale brown
Form
Powder or lumps
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (HPLC)
≥97.5%

Description

It is used in the preparation of hepatitis C virus NS5b RNA polymerase inhibitors. Also used in the determination of cobalt, copper, nickel, and zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of hepatitis C virus NS5b RNA polymerase inhibitors. Also used in the determination of cobalt, copper, nickel, and zinc.

Solubility
Soluble in methanol and dimethyl sulfoxide.

Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text