Thermo Scientific Chemicals

N,N-Dimethylaniline, 99%, Thermo Scientific Chemicals

Catalog number: A11916.0D
10,000 mL, Each
Thermo Scientific Chemicals

N,N-Dimethylaniline, 99%, Thermo Scientific Chemicals

Catalog number: A11916.0D
10,000 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A11916.0D
also known as A11916-0D
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
121-69-7
IUPAC Name
N,N-dimethylaniline
Molecular Formula
C8H11N
InChI Key
JLTDJTHDQAWBAV-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=CC=C1
Identification (FTIR)
Conforms
Appearance (Color)
Clear pale yellow to yellow
Assay (GC)
≥98.5%
Form
Liquid
Refractive Index
1.5555-1.5595 @ 20?C

Description

N,N-Dimethylaniline acts as a precursor to triarylmethane dyes such as crystal violet and malachite green. It is also used in the synthesis of vanillin, methyl violet, and Michler's ketone. Further, it is used as a hardener for plastic resins and an acid scavenger in the manufacture of semisynthetic penicillins and cephalosporins. In addition to this, it is used as a promoter in the curing of polyester and vinyl ester resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylaniline acts as a precursor to triarylmethane dyes such as crystal violet and malachite green. It is also used in the synthesis of vanillin, methyl violet, and Michler′s ketone. Further, it is used as a hardener for plastic resins and an acid scavenger in the manufacture of semisynthetic penicillins and cephalosporins. In addition to this, it is used as a promoter in the curing of polyester and vinyl ester resins.

Solubility
Miscible with water, ethanol, ether, acetone, chloroform, benzene and alkaline.

Notes
Incompatible with strong oxidizing agents, Strong acids, acid chlorides, acid anhydrides, chloroformates and halogens.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text