Thermo Scientific Chemicals

cis-Stilbene, 97%, Thermo Scientific Chemicals

Catalog number: A11924.03
1 g, Each
Thermo Scientific Chemicals

cis-Stilbene, 97%, Thermo Scientific Chemicals

Catalog number: A11924.03
1 g, Each
Quantity
Catalog number: A11924.03
also known as A11924-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
645-49-8
IUPAC Name
[(1Z)-2-phenylethenyl]benzene
Molecular Formula
C14H12
InChI Key
PJANXHGTPQOBST-QXMHVHEDSA-N
SMILES
C(=CC1=CC=CC=C1)C1=CC=CC=C1
Assay (GC)
≥96.0%
Form
Liquid
Refractive Index
1.6185-1.6240 @ 20?C
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow

Description

cis-Stilbene is used as a phosphor and a scintillator. It is mainly used in the produciton of dyes and optical brighteners.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-Stilbene is used as a phosphor and a scintillator. It is mainly used in the produciton of dyes and optical brighteners.

Solubility
Immiscible with water. Miscible with ethanol.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text