cis-Stilbene, 97%, Thermo Scientific Chemicals
cis-Stilbene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

cis-Stilbene, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number A11924.06
also known as A11924-06
Price (USD)/ Each
69.65
Online exclusive
77.50 
Save 7.85 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
69.65
Online exclusive
77.50 
Save 7.85 (10%)
Add to cart
cis-Stilbene, 97%, Thermo Scientific Chemicals
Catalog numberA11924.06
Price (USD)/ Each
69.65
Online exclusive
77.50 
Save 7.85 (10%)
-
Add to cart
Chemical Identifiers
CAS645-49-8
IUPAC Name[(1Z)-2-phenylethenyl]benzene
Molecular FormulaC14H12
InChI KeyPJANXHGTPQOBST-QXMHVHEDSA-N
SMILESC(=CC1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.6185-1.6240 @ 20?C
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
cis-Stilbene is used as a phosphor and a scintillator. It is mainly used in the produciton of dyes and optical brighteners.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-Stilbene is used as a phosphor and a scintillator. It is mainly used in the produciton of dyes and optical brighteners.

Solubility
Immiscible with water. Miscible with ethanol.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Wang, J.; Huang, J.; Du, L.; Lan, Z. Photoinduced Ultrafast Intramolecular Excited-State Energy Transfer in the Silylene-Bridged Biphenyl andStilbene(SBS) System: A Nonadiabatic Dynamics Point of View. J. Phys. Chem. A 2015, 119 (27), 6937-6948.
  2. Fujitsuka, M.; Cho, D. W.; Choi, J.; Tojo, S.; Majima, T. Detection of Structural Changes upon One-Electron Oxidation and Reduction of Stilbene Derivatives by Time-Resolved Resonance Raman Spectroscopy during Pulse Radiolysis and Theoretical Calculations. J. Phys. Chem. A 2015, 119 (26), 6816-6822.