cis-Stilbene is used as a phosphor and a scintillator. It is mainly used in the produciton of dyes and optical brighteners.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
cis-Stilbene is used as a phosphor and a scintillator. It is mainly used in the produciton of dyes and optical brighteners.
Solubility
Immiscible with water. Miscible with ethanol.
Notes
Incompatible with oxidizing agents.
RUO – Research Use Only
Wang, J.; Huang, J.; Du, L.; Lan, Z. Photoinduced Ultrafast Intramolecular Excited-State Energy Transfer in the Silylene-Bridged Biphenyl andStilbene(SBS) System: A Nonadiabatic Dynamics Point of View. J. Phys. Chem. A 2015, 119 (27), 6937-6948.
Fujitsuka, M.; Cho, D. W.; Choi, J.; Tojo, S.; Majima, T. Detection of Structural Changes upon One-Electron Oxidation and Reduction of Stilbene Derivatives by Time-Resolved Resonance Raman Spectroscopy during Pulse Radiolysis and Theoretical Calculations. J. Phys. Chem. A 2015, 119 (26), 6816-6822.