Thermo Scientific Chemicals

Iodic acid, 99%, Thermo Scientific Chemicals

Catalog number: A11925.18
50 g, Each
Thermo Scientific Chemicals

Iodic acid, 99%, Thermo Scientific Chemicals

Catalog number: A11925.18
50 g, Each
Quantity
Catalog number: A11925.18
also known as A11925-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7782-68-5
IUPAC Name
lithium(1+) iodate
Molecular Formula
ILiO3
InChI Key
FZAXZVHFYFGNBX-UHFFFAOYSA-M
SMILES
[Li+].[O-][I](=O)=O
Assay (Iodometric Titration)
≥98.5 to ≤101.5%
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale cream

Description

Iodic acid is used in analytical chemistry laboratories to standardize solutions of both weak and strong bases using methyl red or methyl orange as the indicator. It acts as a key starting material to synthesize sodium or potassium iodate, thereby increasing the iodine content of salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodic acid is used in analytical chemistry laboratories to standardize solutions of both weak and strong bases using methyl red or methyl orange as the indicator. It acts as a key starting material to synthesize sodium or potassium iodate, thereby increasing the iodine content of salt.

Solubility
Soluble in water and methanol.

Notes
Light sensitive. Incompatible with reducing agents, alcohols and organic materials.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text