2'-Methoxyacetophenone, 98%, Thermo Scientific Chemicals
2'-Methoxyacetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2'-Methoxyacetophenone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number A11929.22
also known as A11929-22
Price (USD)/ Each
190.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
190.00
Add to cart
2'-Methoxyacetophenone, 98%, Thermo Scientific Chemicals
Catalog numberA11929.22
Price (USD)/ Each
190.00
-
Add to cart
Chemical Identifiers
CAS579-74-8
IUPAC Name1-(2-methoxyphenyl)ethan-1-one
Molecular FormulaC9H10O2
InChI KeyDWPLEOPKBWNPQV-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5380-1.5420 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only