Thermo Scientific Chemicals

Thiophene-2-carbonitrile, 98%, Thermo Scientific Chemicals

Catalog number: A11930.14
25 g, Each
Thermo Scientific Chemicals

Thiophene-2-carbonitrile, 98%, Thermo Scientific Chemicals

Catalog number: A11930.14
25 g, Each
Quantity
Catalog number: A11930.14
also known as A11930-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1003-31-2
IUPAC Name
thiophene-2-carbonitrile
Molecular Formula
C5H3NS
InChI Key
CUPOOAWTRIURFT-UHFFFAOYSA-N
SMILES
N#CC1=CC=CS1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.5620-1.5670 @ 20°C

Description

Thiophene-2-carbonitrile is used as an intermediate in synthetic chemistry and in the pharmaceutical industry. It is also used in the preparation of thiaplatinacycles and 2,2'-thienylpyrroles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene-2-carbonitrile is used as an intermediate in synthetic chemistry and in the pharmaceutical industry. It is also used in the preparation of thiaplatinacycles and 2,2′-thienylpyrroles.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text