1,3-Diaminopropane, 98%, Thermo Scientific Chemicals
1,3-Diaminopropane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Diaminopropane, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A11932.AE
Price (USD)/ Each
28.65
Online exclusive
31.20 
Save 2.55 (8%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
28.65
Online exclusive
31.20 
Save 2.55 (8%)
Add to cart
1,3-Diaminopropane, 98%, Thermo Scientific Chemicals
Catalog numberA11932.AE
Price (USD)/ Each
28.65
Online exclusive
31.20 
Save 2.55 (8%)
-
Add to cart
Chemical Identifiers
CAS109-76-2
IUPAC Namepropane-1,3-diamine
Molecular FormulaC3H10N2
InChI KeyXFNJVJPLKCPIBV-UHFFFAOYSA-N
SMILESNCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.4555-1.4595 @ 20?C
1,3-Diaminopropane is commonly used in the preparation of cholinesterase inhibitors and thrombosis inhibitors. It is also used as a polymer cross linkage agent, polyurethane extender and spray coatings. It is involved in the preparation of agrochemicals and pharmaceutical intermediates. Further, it acts as a building block in the synthesis of heterocyclic compounds, which finds application in textile finishing. In addition to this, it is used as a ligand and form coordination complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diaminopropane is commonly used in the preparation of cholinesterase inhibitors and thrombosis inhibitors. It is also used as a polymer cross linkage agent, polyurethane extender and spray coatings. It is involved in the preparation of agrochemicals and pharmaceutical intermediates. Further, it acts as a building block in the synthesis of heterocyclic compounds, which finds application in textile finishing. In addition to this, it is used as a ligand and form coordination complexes.

Solubility
Miscible with water.

Notes
Hygroscopic. Air sensitive. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
RUO – Research Use Only

General References:

  1. The potassium derivative (KAPA) is a very strong base, valuable for its ability to cause the migration of acetylenic unsaturation to the terminal positions of alkynes (known as the zip reaction). In the presence of excess amine, reaction occurs within seconds; the driving force is the formation of a stable acetylide anion: J. Am. Chem. Soc., 97, 891 (1975). For example, with acetylenic carbinols: J. Chem. Soc., Chem. Commun., 959 (1976); Tetrahedron Lett., 2565 (1976); Can. J. Chem., 62, 1333 (1984). For a further example (2- to 9-decynol), using a combination of Potassium tert-butoxide, A13947, and Li 3-aminopropanamide, see: Org. Synth. Coll., 8, 146 (1993). This technique avoids the use of KH. For the use of KAPA for the isomerization of ß-pinene to the thermodynamically-more stable ɑ-pinene, see: Org. Synth. Coll., 8, 553 (1993).
  2. For procedures for the generation of Na or K 3-amino-propylamides from the metals by ultrasound irradiation or in the presence of Fe(NO3)3, see: J. Org. Chem., 49, 2494 (1984).
  3. Bag, P.; Ghosh, A. B.; Dutta, A. K.; Flörke, U.; Nag, K. Sandwich-type lanthanide(III) complexes of a tetraiminodiphenol macrocyclic ligand derived from 4-methyl-2,6-diformylphenol and 1,3-diaminopropane: structure, NMR and luminescence. Polyhedron 2015, 102, 539-548.
  4. Liang, K. L.; Lin, Y. F.; Leron, R. B.; Li, M. H. Molar heat capacity of aqueous solutions of 1, 3-diaminopropane and 1, 4-diaminobutane and their piperazine blends. Thermochim. Acta 2015, 616, 42-48.