Thermo Scientific Chemicals

Nitrilotriacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: A11936.22
100 g, Each
Thermo Scientific Chemicals

Nitrilotriacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: A11936.22
100 g, Each
Quantity
Catalog number: A11936.22
also known as A11936-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
139-13-9
IUPAC Name
2-[bis(carboxymethyl)amino]acetic acid
Molecular Formula
C6H9NO6
InChI Key
MGFYIUFZLHCRTH-UHFFFAOYSA-N
SMILES
OC(=O)CN(CC(O)=O)CC(O)=O
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

It is employed as an excellent chelating agent. It is also used as a complexing agent, metal masking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an excellent chelating agent. It is also used as a complexing agent, metal masking agent.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text