Thermo Scientific Chemicals

3-Bromobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A11941.14
25 g, Each
Thermo Scientific Chemicals

3-Bromobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A11941.14
25 g, Each
Quantity
Catalog number: A11941.14
also known as A11941-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3132-99-8
IUPAC Name
3-bromobenzaldehyde
Molecular Formula
C7H5BrO
InChI Key
SUISZCALMBHJQX-UHFFFAOYSA-N
SMILES
BrC1=CC=CC(C=O)=C1
Appearance (Color)
Clear colorless to pale yellow
Free acid (titration)
<1.5%
Assay (GC)
≥96.0%
Refractive Index
1.5915-1.5965 @ 20?C
Form
Liquid

Description

A substituted benzaldehyde used in the synthesis of more complex pharmaceutical compounds. It is also employed as an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A substituted benzaldehyde used in the synthesis of more complex pharmaceutical compounds. It is also employed as an active pharmaceutical ingredient.

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Keep away from oxidizing agents and air. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text