3-Aminophenol, 98+%, Thermo Scientific Chemicals
3-Aminophenol, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Aminophenol, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
250 g
5000 g
Catalog number A11943.0B
also known as A11943-0B
Price (USD)/ Each
339.00
-
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)/ Each
339.00
Add to cart
3-Aminophenol, 98+%, Thermo Scientific Chemicals
Catalog numberA11943.0B
Price (USD)/ Each
339.00
-
Add to cart
Chemical Identifiers
CAS591-27-5
IUPAC Name3-aminophenol
Molecular FormulaC6H7NO
InChI KeyCWLKGDAVCFYWJK-UHFFFAOYSA-N
SMILESNC1=CC=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown or grey
Assay (GC)≥98.0%
Melting Point (clear melt)119.5-125.5?C
FormCrystals or powder or crystalline powder or flakes
Identification (FTIR)Conforms
A chemical used in the preparation of inhibitors of mammalian carbonic anhydrase isoforms. It is also used in the preparation of quinoline tethered fluorescent carbon dots for regulated anticancer discovery. It is employed in the production of TB drugs, salicylic acid, amino stabilizer and developers. 3-Aminophenol has been used as a stabilizer of chlorine-containing thermoplastics, although its principal use is as an intermediate in the production of 4-amino-2-hydroxybenzoic acid this isomer is also employed as a hair colorant and as a coupler molecule in hair dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A chemical used in the preparation of inhibitors of mammalian carbonic anhydrase isoforms. It is also used in the preparation of quinoline tethered fluorescent carbon dots for regulated anticancer discovery. It is employed in the production of TB drugs, salicylic acid, amino stabilizer and developers. 3-Aminophenol has been used as a stabilizer of chlorine-containing thermoplastics, although its principal use is as an intermediate in the production of 4-amino-2-hydroxybenzoic acid this isomer is also employed as a hair colorant and as a coupler molecule in hair dyes.

Solubility
Soluble in water (35 mg/ml at 20°C), ether, alcohol, amyl alcohol, and benzene (slightly).

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
RUO – Research Use Only

General References:

  1. P. Nagaraja.; H.S. Yathirajan.; C.R. Raju.; R.A. Vasantha.; P. Nagendra.; M.S. Hemantha Kumar. 3-Aminophenol as a novel coupling agent for the spectrophotometric determination of sulfonamide derivatives.Il Farmaco,2003,58(12), 1295-1300.
  2. Padmarajaiah Nagaraja.; Ramanathapura A Vasantha.; Hemmige S Yathirajan. Spectrophotometric determination of folic acid in pharmaceutical preparations by coupling reactions with iminodibenzyl or 3-aminophenol or sodium molybdate-pyrocatechol.Anal. Biochem.2002,307(2), 316-321.