Thermo Scientific Chemicals

Antimony(III) chloride, 99+%, Thermo Scientific Chemicals

Catalog number: A11944.22
100 g, Each
Thermo Scientific Chemicals

Antimony(III) chloride, 99+%, Thermo Scientific Chemicals

Catalog number: A11944.22
100 g, Each
Quantity
Catalog number: A11944.22
also known as A11944-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10025-91-9
IUPAC Name
antimony(3+) trichloride
Molecular Formula
Cl3Sb
InChI Key
FAPDDOBMIUGHIN-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Sb+3]
Form
Crystals or powder or crystalline powder or lumps
Assay (Redox Titration)
≥99.0 to ≤101.0%
Appearance (Color)
White to brown

Description

Antimony(III) chloride is engaged to detect certain compounds, such as cholesterols. It is a complexing agent used in the colorimetric Carr-Price test for detecting carotenoids including vitamin A. It is employed as a mordant and a fabric flame retardant. It is used as a catalyst for chlorinationn and polymerization reactions. It is used to coat the steel to make bronze, used as a cutting material of antimony salt and as a preservative in medicine. It is used as an analytical reagent for chloral and aromatics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Antimony(III) chloride is engaged to detect certain compounds, such as cholesterols. It is a complexing agent used in the colorimetric Carr-Price test for detecting carotenoids including vitamin A. It is employed as a mordant and a fabric flame retardant. It is used as a catalyst for chlorinationn and polymerization reactions. It is used to coat the steel to make bronze, used as a cutting material of antimony salt and as a preservative in medicine. It is used as an analytical reagent for chloral and aromatics.

Solubility
Soluble in water, hydrochloric acid, alcohol, benzene, carbon disulfide, dioxane, chloroform, ether, acetone and carbon tetrachloride. Insoluble in organic bases.

Notes
Moisture sensitive and hygroscopic in nature. It is corrosive to metals and tissue. Incompatible with strong oxidizing agents, strong acids and strong bases. Decomposes in water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text