Thermo Scientific Chemicals

4-Fluorophenol, 99%, Thermo Scientific Chemicals

Catalog number: A11945.14
25 g, Each
Thermo Scientific Chemicals

4-Fluorophenol, 99%, Thermo Scientific Chemicals

Catalog number: A11945.14
25 g, Each
Quantity
Catalog number: A11945.14
also known as A11945-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
371-41-5
IUPAC Name
4-fluorophenol
Molecular Formula
C6H5FO
InChI Key
RHMPLDJJXGPMEX-UHFFFAOYSA-N
SMILES
OC1=CC=C(F)C=C1
Assay (GC)
≥98.5%
Melting Point (clear melt)
43.0-51.0?C
Form
Crystals or powder or crystalline powder or lumps or fused solid
Appearance (Color)
White to pale cream to cream to yellow
Identification (FTIR)
Conforms

Description

4-Fluorophenol is a fluorinated phenolic compound with various applications as an starting reagent for the synthesis of pharmaceutical goods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophenol is a fluorinated phenolic compound with various applications as an starting reagent for the synthesis of pharmaceutical goods.

Solubility
Soluble in water (80 mg/ml at 20°C), methylene dichloride, chloroform and methanol.

Notes
Keep container tightly closed in a dry and well-ventilated place. Store away from strong bases and oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text