Thermo Scientific Chemicals

o-Phenylenediamine, 98%, Thermo Scientific Chemicals

Catalog number: A11946.14
25 g, Each
Thermo Scientific Chemicals

o-Phenylenediamine, 98%, Thermo Scientific Chemicals

Catalog number: A11946.14
25 g, Each
Quantity
Catalog number: A11946.14
also known as A11946-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95-54-5
IUPAC Name
benzene-1,2-diamine
Molecular Formula
C6H8N2
InChI Key
GEYOCULIXLDCMW-UHFFFAOYSA-N
SMILES
NC1=CC=CC=C1N
Appearance (Color)
White to pale brown
Assay (GC)
≥97.5%
Melting Point (clear melt)
99-104?C
Form
Crystals, flakes or pellets
Identification (FTIR)
Conforms

Description

o-phenylenediamine is used to prepare tiabendazole, pyrazinamide, morinamide, clemizole and chlormidazole. It is involved in the synthesis of benzimidazole by reacting with carboxylic acids and their derivatives. It is also used to prepare quinoxalinedione and mercaptoimidazoles by condensation with dimethyl oxalate and xanthate esters respectively. It reacts with nitrous acid to get benzotriazole, which finds application as a corrosion inhibitor. It is actively involved in the preparation of pharmaceuticals by reacting substituted o-phenylenediamine with various diketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-phenylenediamine is used to prepare tiabendazole, pyrazinamide, morinamide, clemizole and chlormidazole. It is involved in the synthesis of benzimidazole by reacting with carboxylic acids and their derivatives. It is also used to prepare quinoxalinedione and mercaptoimidazoles by condensation with dimethyl oxalate and xanthate esters respectively. It reacts with nitrous acid to get benzotriazole, which finds application as a corrosion inhibitor. It is actively involved in the preparation of pharmaceuticals by reacting substituted o-phenylenediamine with various diketones.

Solubility
Soluble in alcohol, chloroform, ether, slightly soluble in water. Soluble in hot water.

Notes
Moisture and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text