Cyclopropanecarbonyl chloride, 98%, Thermo Scientific Chemicals
Cyclopropanecarbonyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopropanecarbonyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A11948.14
also known as A11948-14
Price (USD)/ Each
46.80
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
46.80
Add to cart
Cyclopropanecarbonyl chloride, 98%, Thermo Scientific Chemicals
Catalog numberA11948.14
Price (USD)/ Each
46.80
-
Add to cart
Chemical Identifiers
CAS4023-34-1
IUPAC Namecyclopropanecarbonyl chloride
Molecular FormulaC4H5ClO
InChI KeyZOOSILUVXHVRJE-UHFFFAOYSA-N
SMILESClC(=O)C1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.4510-1.4550 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
FormLiquid
It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.

Solubility
Soluble in water (reacts violently).

Notes
Moisture Sensitive. Store away from oxidizing agents, bases, active metals and water/moisture. Store under inert gas.
RUO – Research Use Only

General References:

  1. J. S. A. M. de Boer and C. H. Stam. The influence of substituents on the geometry of the cyclopropane ring. VI The molecular and crystal structures of cyclopropanecarboxylic acid and cyclopropanecarbonyl chloride.Recl. Trav. Chim. Pays-Bas.1992,111(9), 407-410.
  2. Yung-Lung Lin.; Chung-Shieh Wu.; Shean-Woei Lin.; Jian-Lin Huang.; Yang-Sheng Sun.; Ding-Yah Yang. SAR Studies of 3-Cyclopropanecarbonyloxy-2-cyclohexen-1-one as Inhibitors of 4-Hydroxyphenylpyruvate Dioxygenase.Bioorg. Med. Chem.2002,10(3), 685-690.