4-Isopropylbenzonitrile, 95%, Thermo Scientific Chemicals
4-Isopropylbenzonitrile, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Isopropylbenzonitrile, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A11954.03
also known as A11954-03
Price (USD)/ Each
45.50
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
45.50
Add to cart
4-Isopropylbenzonitrile, 95%, Thermo Scientific Chemicals
Catalog numberA11954.03
Price (USD)/ Each
45.50
-
Add to cart
Chemical Identifiers
CAS13816-33-6
IUPAC Name4-(propan-2-yl)benzonitrile
Molecular FormulaC10H11N
InChI KeyYFDJCWXBKWRDPW-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥94.0%
Refractive Index1.5145-1.5205 @ 20°C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only