Thermo Scientific Chemicals

Starch, from potato, soluble

Catalog number: A11961.36
500 g, Each
Thermo Scientific Chemicals

Starch, from potato, soluble

Catalog number: A11961.36
500 g, Each
Quantity
Catalog number: A11961.36
also known as A11961-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9005-84-9
IUPAC Name
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Molecular Formula
C12H22O11
InChI Key
GUBGYTABKSRVRQ-ASMJPISFSA-N
SMILES
OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
pH
5.0 - 7.0 (1%, water)
Appearance (Color)
White
Form
Powder
Loss on Drying
≤ 20%

Description

In food industry, starch is used as a food preservative. Starch has extensive industrial use as an adhesive or bonding agent. It is also used in paper industry. Starch is used in the procedure to manufacture gypsum wallboards. Starch used in the laundry, also known as clothing starch. Glucose derived from starch may be fermented further to make ethanol or biofuel. Starch is also employed in the petroleum exploration industry. It is interesting to note that starch may be used to make hydrogen by making usage of enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text