2,6-Dichlorophenylhydrazine hydrochloride, 98+%, Thermo Scientific Chemicals
2,6-Dichlorophenylhydrazine hydrochloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Dichlorophenylhydrazine hydrochloride, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number A11968.06
also known as A11968-06
Price (USD)/ Each
99.60
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
99.60
Add to cart
2,6-Dichlorophenylhydrazine hydrochloride, 98+%, Thermo Scientific Chemicals
Catalog numberA11968.06
Price (USD)/ Each
99.60
-
Add to cart
Chemical Identifiers
CAS50709-36-9
IUPAC Namehydrogen (2,6-dichlorophenyl)hydrazine chloride
Molecular FormulaC6H7Cl3N2
InChI KeyCQNIYLLTIOPFCJ-UHFFFAOYSA-N
SMILES[H+].[Cl-].NNC1=C(Cl)C=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale yellow
FormPowder
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only