(+/-)-Phenylsuccinic acid, 98%, Thermo Scientific Chemicals
(+/-)-Phenylsuccinic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(+/-)-Phenylsuccinic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A11971.14
also known as A11971-14
Price (USD)/ Each
29.20
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
29.20
Add to cart
(+/-)-Phenylsuccinic acid, 98%, Thermo Scientific Chemicals
Catalog numberA11971.14
Price (USD)/ Each
29.20
-
Add to cart
Chemical Identifiers
CAS635-51-8
IUPAC Name2-phenylbutanedioic acid
Molecular FormulaC10H10O4
InChI KeyLVFFZQQWIZURIO-UHFFFAOYNA-N
SMILESOC(=O)CC(C(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Formpowder
Assay (Aqueous acid-base Titration)>97.5%
Assay (Silylated GC)>97.5%
Water Content (Karl Fischer Titration)<0.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only