Thermo Scientific Chemicals

12-Crown-4, 98%, Thermo Scientific Chemicals

Catalog number: A11972.14
25 g, Each
Thermo Scientific Chemicals

12-Crown-4, 98%, Thermo Scientific Chemicals

Catalog number: A11972.14
25 g, Each
Quantity
Catalog number: A11972.14
also known as A11972-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
294-93-9
IUPAC Name
1,4,7,10-tetraoxacyclododecane
Molecular Formula
C8H16O4
InChI Key
XQQZRZQVBFHBHL-UHFFFAOYSA-N
SMILES
C1COCCOCCOCCO1
Refractive Index
1.4600-1.4640 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms

Description

It serves as an aprotic solvent for alkali metals, the sodium solution is a strong reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It serves as an aprotic “solvent” for alkali metals, the sodium solution is a strong reducing agent.

Solubility
Fully miscible in water.

Notes
Light & Air Sensitive & Hygroscopic. Store under inert gas. Keep in a tightly closed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text