Thermo Scientific Chemicals

1-Nitropropane, 98%, Thermo Scientific Chemicals

Catalog number: A11975.36
500 g, Each
Thermo Scientific Chemicals

1-Nitropropane, 98%, Thermo Scientific Chemicals

Catalog number: A11975.36
500 g, Each
Quantity
Catalog number: A11975.36
also known as A11975-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-03-2
IUPAC Name
1-nitropropane
Molecular Formula
C3H7NO2
InChI Key
JSZOAYXJRCEYSX-UHFFFAOYSA-N
SMILES
CCC[N+]([O-])=O
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.4000-1.4040 @ 20?C

Description

The solvent of 1-Nitropropane is mainly used coatings, inks and separation processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The solvent of 1-Nitropropane is mainly used coatings, inks and separation processes.

Solubility
Soluble in water. (1.4 g/L)

Notes
Store in cool place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text