Thermo Scientific Chemicals

Veratrole, 99%, Thermo Scientific Chemicals

Catalog number: A11985.0B
1000 g, Each
Thermo Scientific Chemicals

Veratrole, 99%, Thermo Scientific Chemicals

Catalog number: A11985.0B
1000 g, Each
Quantity
Catalog number: A11985.0B
also known as A11985-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
91-16-7
IUPAC Name
1,2-dimethoxybenzene
Molecular Formula
C8H10O2
InChI Key
ABDKAPXRBAPSQN-UHFFFAOYSA-N
SMILES
COC1=CC=CC=C1OC
Form
Liquid as melt
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.5320-1.5350 @ 20?C
Appearance (Color)
Clear colorless to pale yellow

Description

It is useful for organic synthesis of aromatic compounds. Veratrole is relatively electron-rich and thus readily undergoes electrophilic substitution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is useful for organic synthesis of aromatic compounds. Veratrole is relatively electron-rich and thus readily undergoes electrophilic substitution.

Solubility
Soluble in alcohol, diethyl ether, acetone, and methanol. Slightly soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text